Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601993 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCOA2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NCOA2 - nuclear receptor coactivator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321703.1 | 5012 | UTR 3 | NP_001308632.1 | |||
NM_001321707.1 | 5012 | UTR 3 | NP_001308636.1 | |||
NM_001321711.1 | 5012 | UTR 3 | NP_001308640.1 | |||
NM_001321712.1 | 5012 | UTR 3 | NP_001308641.1 | |||
NM_001321713.1 | 5012 | UTR 3 | NP_001308642.1 | |||
NM_006540.3 | 5012 | UTR 3 | NP_006531.1 | |||
XM_017012961.1 | 5012 | UTR 3 | XP_016868450.1 | |||
XM_017012962.1 | 5012 | UTR 3 | XP_016868451.1 | |||
XM_017012963.1 | 5012 | UTR 3 | XP_016868452.1 | |||
XM_017012964.1 | 5012 | UTR 3 | XP_016868453.1 | |||
XM_017012965.1 | 5012 | UTR 3 | XP_016868454.1 | |||
XM_017012966.1 | 5012 | UTR 3 | XP_016868455.1 | |||
XM_017012967.1 | 5012 | UTR 3 | XP_016868456.1 | |||
XM_017012968.1 | 5012 | UTR 3 | XP_016868457.1 | |||
XM_017012969.1 | 5012 | UTR 3 | XP_016868458.1 | |||
XM_017012970.1 | 5012 | UTR 3 | XP_016868459.1 | |||
XM_017012971.1 | 5012 | UTR 3 | XP_016868460.1 | |||
XM_017012972.1 | 5012 | UTR 3 | XP_016868461.1 | |||
XM_017012973.1 | 5012 | Intron | XP_016868462.1 |
Set Membership: |
HapMap |