Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGGCAGCCCCGCTTGCTGGCAGC[A/G]GTGGCTGGGGTTTCCTTCCCATAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608863 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDPN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDPN - podoplanin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006624.1 | Intron | NP_001006625.1 | ||||
NM_001006625.1 | Intron | NP_001006626.1 | ||||
NM_006474.4 | Intron | NP_006465.3 | ||||
NM_198389.2 | Intron | NP_938203.2 | ||||
XM_006710295.1 | Intron | XP_006710358.1 |