Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606122 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNFRSF19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNFRSF19 - TNF receptor superfamily member 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204458.1 | Intron | NP_001191387.1 | ||||
NM_001204459.1 | Intron | NP_001191388.1 | ||||
NM_018647.3 | Intron | NP_061117.2 | ||||
NM_148957.3 | Intron | NP_683760.1 | ||||
XM_005266445.2 | Intron | XP_005266502.1 | ||||
XM_005266446.1 | Intron | XP_005266503.1 | ||||
XM_011535146.1 | Intron | XP_011533448.1 | ||||
XM_017020651.1 | Intron | XP_016876140.1 | ||||
XM_017020652.1 | Intron | XP_016876141.1 |