Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf118 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C6orf118 - chromosome 6 open reading frame 118 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144980.3 | Intron | NP_659417.2 | ||||
XM_005266838.2 | Intron | XP_005266895.1 | ||||
XM_011535509.2 | Intron | XP_011533811.1 | ||||
XM_011535510.2 | Intron | XP_011533812.1 | ||||
XM_011535511.2 | Intron | XP_011533813.1 | ||||
XM_011535512.2 | Intron | XP_011533814.1 | ||||
XM_011535513.2 | Intron | XP_011533815.1 | ||||
XM_017010323.1 | Intron | XP_016865812.1 | ||||
XM_017010324.1 | Intron | XP_016865813.1 |