Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACGCCTCCAATCCCAGGTACTGC[C/T]CACCTTAAGTCCTCCCTGCCTGCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603470 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ASS1 - argininosuccinate synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000050.4 | Intron | NP_000041.2 | ||||
NM_054012.3 | Intron | NP_446464.1 | ||||
XM_005272200.3 | Intron | XP_005272257.1 | ||||
XM_011518705.2 | Intron | XP_011517007.1 | ||||
XM_017014729.1 | Intron | XP_016870218.1 |
Set Membership: |
HapMap |