Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAATCATAAAATGTATAGTGAC[C/T]GTACTGATTCATATCAGTCTTTAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603803 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DACH1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DACH1 - dachshund family transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004392.6 | 3003 | UTR 3 | NP_004383.4 | |||
NM_080759.5 | 3003 | UTR 3 | NP_542937.3 | |||
NM_080760.5 | 3003 | UTR 3 | NP_542938.3 | |||
XM_011534939.2 | 3003 | UTR 3 | XP_011533241.1 | |||
XM_011534940.2 | 3003 | Intron | XP_011533242.1 | |||
XM_011534941.2 | 3003 | Intron | XP_011533243.1 | |||
XM_011534942.2 | 3003 | UTR 3 | XP_011533244.1 | |||
XM_017020396.1 | 3003 | UTR 3 | XP_016875885.1 |
Set Membership: |
HapMap |