Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609042 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OPN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
OPN5 - opsin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181744.3 | 79 | Intron | NP_859528.1 | |||
XM_017010409.1 | 79 | Intron | XP_016865898.1 | |||
XM_017010410.1 | 79 | Intron | XP_016865899.1 | |||
XM_017010411.1 | 79 | Intron | XP_016865900.1 | |||
XM_017010412.1 | 79 | Intron | XP_016865901.1 | |||
XM_017010413.1 | 79 | UTR 5 | XP_016865902.1 | |||
XM_017010414.1 | 79 | Intron | XP_016865903.1 | |||
XM_017010415.1 | 79 | Intron | XP_016865904.1 | |||
XM_017010416.1 | 79 | Intron | XP_016865905.1 |
Set Membership: |
HapMap Validated |