Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610335 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF20 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PHF20 - PHD finger protein 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016436.4 | Intron | NP_057520.2 | ||||
XM_005260421.2 | Intron | XP_005260478.1 | ||||
XM_011528843.1 | Intron | XP_011527145.1 | ||||
XM_011528844.1 | Intron | XP_011527146.1 | ||||
XM_017027864.1 | Intron | XP_016883353.1 | ||||
XM_017027865.1 | Intron | XP_016883354.1 | ||||
XM_017027866.1 | Intron | XP_016883355.1 | ||||
XM_017027867.1 | Intron | XP_016883356.1 | ||||
XM_017027868.1 | Intron | XP_016883357.1 | ||||
XM_017027869.1 | Intron | XP_016883358.1 | ||||
XM_017027870.1 | Intron | XP_016883359.1 | ||||
XM_017027871.1 | Intron | XP_016883360.1 |