Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613597 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOGA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HOGA1 - 4-hydroxy-2-oxoglutarate aldolase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MORN4 - MORN repeat containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098831.1 | 674 | Intron | NP_001092301.1 | |||
NM_178832.3 | 674 | Silent Mutation | CGA,CGG | R,R 123 | NP_849154.1 | |
XM_011539250.2 | 674 | Silent Mutation | CGA,CGG | R,R 181 | XP_011537552.1 | |
XM_011539251.2 | 674 | Silent Mutation | CGA,CGG | R,R 123 | XP_011537553.1 |