Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGAGCACGGGGGCCACCTGTGG[A/G]AGGGTGGAGAGAGATTCCAGCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616569 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSAD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CSAD - cysteine sulfinic acid decarboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244705.1 | Intron | NP_001231634.1 | ||||
NM_001244706.1 | Intron | NP_001231635.1 | ||||
NM_015989.4 | Intron | NP_057073.4 | ||||
XM_011538442.2 | Intron | XP_011536744.2 | ||||
XM_011538446.2 | Intron | XP_011536748.1 | ||||
XM_011538449.1 | Intron | XP_011536751.1 | ||||
XM_011538451.1 | Intron | XP_011536753.1 | ||||
XM_017019416.1 | Intron | XP_016874905.1 | ||||
XM_017019417.1 | Intron | XP_016874906.1 | ||||
XM_017019418.1 | Intron | XP_016874907.1 | ||||
XM_017019419.1 | Intron | XP_016874908.1 |
Set Membership: |
HapMap Validated |