Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607717 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6757 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MIR6757 - microRNA 6757 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPRYD3 - SPRY domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032840.2 | 2030 | UTR 3 | NP_116229.1 |
TNS2 - tensin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015319.2 | 2030 | Intron | NP_056134.2 | |||
NM_170754.2 | 2030 | Intron | NP_736610.2 | |||
NM_198316.1 | 2030 | Intron | NP_938072.1 | |||
XM_006719302.3 | 2030 | Intron | XP_006719365.1 | |||
XM_006719303.1 | 2030 | Intron | XP_006719366.1 | |||
XM_011538079.1 | 2030 | Intron | XP_011536381.1 | |||
XM_017019088.1 | 2030 | Intron | XP_016874577.1 | |||
XM_017019089.1 | 2030 | Intron | XP_016874578.1 |
Set Membership: |
HapMap Validated |