Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCGAGTGCCACAGGATGGGAGGG[A/G]CTCACAGCCACAGTGGATCCTGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603928 MIM: 607717 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF4B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EIF4B - eukaryotic translation initiation factor 4B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC283335 - uncharacterized LOC283335 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6757 - microRNA 6757 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNS2 - tensin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015319.2 | 617 | Intron | NP_056134.2 | |||
NM_170754.2 | 617 | Intron | NP_736610.2 | |||
NM_198316.1 | 617 | Intron | NP_938072.1 | |||
XM_006719302.3 | 617 | UTR 5 | XP_006719365.1 | |||
XM_006719303.1 | 617 | Intron | XP_006719366.1 | |||
XM_011538079.1 | 617 | Intron | XP_011536381.1 | |||
XM_017019088.1 | 617 | Intron | XP_016874577.1 | |||
XM_017019089.1 | 617 | Intron | XP_016874578.1 |
Set Membership: |
Validated |