Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102582 MIM: 601511 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STAT3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
STAT3 - signal transducer and activator of transcription 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003150.3 | 4199 | UTR 3 | NP_003141.2 | |||
NM_139276.2 | 4199 | UTR 3 | NP_644805.1 | |||
NM_213662.1 | 4199 | UTR 3 | NP_998827.1 | |||
XM_005257616.3 | 4199 | Intron | XP_005257673.2 | |||
XM_005257617.3 | 4199 | Intron | XP_005257674.2 | |||
XM_011525145.2 | 4199 | UTR 3 | XP_011523447.1 | |||
XM_011525146.2 | 4199 | UTR 3 | XP_011523448.1 | |||
XM_017024972.1 | 4199 | Intron | XP_016880461.1 | |||
XM_017024973.1 | 4199 | UTR 3 | XP_016880462.1 | |||
XM_017024974.1 | 4199 | Intron | XP_016880463.1 | |||
XM_017024975.1 | 4199 | Intron | XP_016880464.1 | |||
XM_017024976.1 | 4199 | Intron | XP_016880465.1 |
STAT5A - signal transducer and activator of transcription 5A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |