Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTTTGGAAAATGACATGAAACA[C/T]GAAACATGTTTAAAAACACAATCAA
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 616379 | |||||||||||||||||||||||||||||
Literature Links: |
UBXN7 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
UBXN7 - UBX domain protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015562.1 | 9432 | Intron | NP_056377.1 | |||
XM_011512670.2 | 9432 | UTR 3 | XP_011510972.1 | |||
XM_011512671.2 | 9432 | UTR 3 | XP_011510973.1 |