Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCTGGTTTTTGGAGTCCCGAGCT[A/G]GTGGCCAGCTCTCTCAGCCCCACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605431 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAPK8IP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MAPK8IP3 - mitogen-activated protein kinase 8 interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040439.1 | Intron | NP_001035529.1 | ||||
NM_001318852.1 | Intron | NP_001305781.1 | ||||
NM_015133.4 | Intron | NP_055948.2 | ||||
XM_005255190.2 | Intron | XP_005255247.1 | ||||
XM_006720869.2 | Intron | XP_006720932.1 | ||||
XM_011522429.2 | Intron | XP_011520731.1 | ||||
XM_011522430.2 | Intron | XP_011520732.1 | ||||
XM_011522431.2 | Intron | XP_011520733.1 | ||||
XM_011522432.2 | Intron | XP_011520734.1 | ||||
XM_011522433.2 | Intron | XP_011520735.1 | ||||
XM_017023079.1 | Intron | XP_016878568.1 | ||||
XM_017023080.1 | Intron | XP_016878569.1 | ||||
XM_017023081.1 | Intron | XP_016878570.1 |
Set Membership: |
HapMap Validated |