Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
77 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615463 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HYI PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HYI - hydroxypyruvate isomerase (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190880.2 | 1366 | Intron | NP_001177809.1 | |||
NM_001243526.1 | 1366 | Intron | NP_001230455.1 | |||
NM_031207.5 | 1366 | Intron | NP_112484.3 | |||
XM_005271239.4 | 1366 | Intron | XP_005271296.1 | |||
XM_005271240.4 | 1366 | Intron | XP_005271297.1 | |||
XM_006710937.3 | 1366 | UTR 3 | XP_006711000.1 | |||
XM_011542222.2 | 1366 | Intron | XP_011540524.1 |
MIR6735 - microRNA 6735 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SZT2 - seizure threshold 2 homolog (mouse) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015284.3 | 1366 | UTR 3 | NP_056099.3 | |||
XM_005270686.3 | 1366 | UTR 3 | XP_005270743.1 | |||
XM_006710501.3 | 1366 | UTR 3 | XP_006710564.1 | |||
XM_011541106.2 | 1366 | UTR 3 | XP_011539408.1 | |||
XM_011541107.2 | 1366 | UTR 3 | XP_011539409.1 | |||
XM_017000819.1 | 1366 | UTR 3 | XP_016856308.1 | |||
XM_017000820.1 | 1366 | UTR 3 | XP_016856309.1 | |||
XM_017000821.1 | 1366 | UTR 3 | XP_016856310.1 |
Set Membership: |
HapMap JSNP Validated |