Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAATTAGGTTATGCATTTAACAG[T/G]TTCCACTGTACTACTCAATTGACCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602953 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEY1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HEY1 - hes related family bHLH transcription factor with YRPW motif 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040708.1 | 1751 | UTR 3 | NP_001035798.1 | |||
NM_001282851.1 | 1751 | UTR 3 | NP_001269780.1 | |||
NM_012258.3 | 1751 | UTR 3 | NP_036390.3 |
LINC01607 - long intergenic non-protein coding RNA 1607 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101927040 - uncharacterized LOC101927040 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |