Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608141 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP43 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NUP43 - nucleoporin 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198887.2 | 1025 | Intron | NP_942590.1 | |||
XM_005266960.4 | 1025 | Intron | XP_005267017.1 | |||
XM_005266961.3 | 1025 | Intron | XP_005267018.1 | |||
XM_005266962.3 | 1025 | UTR 3 | XP_005267019.1 | |||
XM_011535798.2 | 1025 | Intron | XP_011534100.1 | |||
XM_011535799.2 | 1025 | Intron | XP_011534101.1 |