Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
51 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609853 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPCS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPCS - phosphopantothenoylcysteine synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077447.2 | 895 | UTR 3 | NP_001070915.1 | |||
NM_001287506.1 | 895 | UTR 3 | NP_001274435.1 | |||
NM_001287507.1 | 895 | UTR 3 | NP_001274436.1 | |||
NM_001287508.1 | 895 | UTR 3 | NP_001274437.1 | |||
NM_001287509.1 | 895 | UTR 3 | NP_001274438.1 | |||
NM_001287510.1 | 895 | UTR 3 | NP_001274439.1 | |||
NM_001287511.1 | 895 | Intron | NP_001274440.1 | |||
NM_024664.3 | 895 | UTR 3 | NP_078940.2 |
ZMYND12 - zinc finger MYND-type containing 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |