Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTTCATGAATTCTTTTTTTACA[A/G]TGGCCCTTATGCTGGATTCATTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602589 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FUT8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FUT8 - fucosyltransferase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004480.4 | Intron | NP_004471.4 | ||||
NM_178155.2 | Intron | NP_835368.1 | ||||
NM_178156.2 | Intron | NP_835369.1 | ||||
XM_011536614.2 | Intron | XP_011534916.1 | ||||
XM_017021136.1 | Intron | XP_016876625.1 | ||||
XM_017021137.1 | Intron | XP_016876626.1 | ||||
XM_017021138.1 | Intron | XP_016876627.1 | ||||
XM_017021139.1 | Intron | XP_016876628.1 | ||||
XM_017021140.1 | Intron | XP_016876629.1 |
Set Membership: |
HapMap |