Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAATGAACAGCCCTGTACATACAG[A/G]TTTGTTGTTTTGTGTGTGTGTGTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610785 | ||||||||||||||||||||
Literature Links: |
PDIK1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDIK1L - PDLIM1 interacting kinase 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243532.1 | 167 | Intron | NP_001230461.1 | |||
NM_001243533.1 | 167 | Intron | NP_001230462.1 | |||
NM_152835.4 | 167 | Intron | NP_690048.1 | |||
XM_005245743.4 | 167 | Intron | XP_005245800.1 | |||
XM_006710381.3 | 167 | UTR 5 | XP_006710444.1 | |||
XM_011540792.2 | 167 | Intron | XP_011539094.1 |