Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACACAGGGTCCCTTGGGGGGCCT[A/G]AGAGCAGCCATGAGGAGGCACAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615955 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC183 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC183 - coiled-coil domain containing 183 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039374.4 | 51 | UTR 5 | NP_001034463.4 |
CCDC183-AS1 - CCDC183 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM141 - transmembrane protein 141 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |