Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCAGTCACTTTCAGTAGACAATG[C/T]CTTTCACAGATAATGTGAACTTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610025 | ||||||||||||||||||||
Literature Links: |
COL24A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL24A1 - collagen type XXIV alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152890.5 | Intron | NP_690850.2 | ||||
XM_017000924.1 | Intron | XP_016856413.1 | ||||
XM_017000925.1 | Intron | XP_016856414.1 | ||||
XM_017000926.1 | Intron | XP_016856415.1 | ||||
XM_017000927.1 | Intron | XP_016856416.1 | ||||
XM_017000928.1 | Intron | XP_016856417.1 | ||||
XM_017000929.1 | Intron | XP_016856418.1 | ||||
XM_017000930.1 | Intron | XP_016856419.1 |