Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTGGTATCGTATCCCTGTTACT[A/T]CTTCTATTGTGATACCTGTTGGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNJ PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
CCNJ - cyclin J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134375.1 | Intron | NP_001127847.1 | ||||
NM_001134376.1 | Intron | NP_001127848.1 | ||||
NM_019084.4 | Intron | NP_061957.2 | ||||
XM_005269921.3 | Intron | XP_005269978.1 | ||||
XM_011539885.2 | Intron | XP_011538187.1 | ||||
XM_017016354.1 | Intron | XP_016871843.1 | ||||
XM_017016355.1 | Intron | XP_016871844.1 | ||||
XM_017016356.1 | Intron | XP_016871845.1 |
ENTPD1-AS1 - ENTPD1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |