Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAATTCTACTAACAAATATATTT[A/T]CTCTAAAACTAATTTTTCCAAACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612987 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATE1-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATE1-AS1 - ATE1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NSMCE4A - NSE4 homolog A, SMC5-SMC6 complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167865.1 | Intron | NP_001161337.1 | ||||
NM_017615.2 | Intron | NP_060085.2 | ||||
XM_005269929.4 | Intron | XP_005269986.1 | ||||
XM_005269930.3 | Intron | XP_005269987.1 | ||||
XM_011539910.2 | Intron | XP_011538212.1 | ||||
XM_017016359.1 | Intron | XP_016871848.1 |
Set Membership: |
HapMap |