Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607347 MIM: 610851 MIM: 609028 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALPK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALPK1 - alpha kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
AP1AR - adaptor related protein complex 1 associated regulatory protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIFA - TRAF interacting protein with forkhead associated domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052864.2 | 1050 | UTR 3 | NP_443096.1 |