Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGGCCCACCTGCTCCAGGCTGA[C/T]CGCTGCAGCATGTTCCTGTGCCGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600827 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE6C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PDE6C - phosphodiesterase 6C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006204.3 | 420 | Silent Mutation | GAC,GAT | D,D 94 | NP_006195.3 | |
XM_017016322.1 | 420 | UTR 5 | XP_016871811.1 |
Set Membership: |
HapMap |