Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602897 MIM: 600267 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAPK10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MAPK10 - mitogen-activated protein kinase 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTPN13 - protein tyrosine phosphatase, non-receptor type 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006264.2 | Intron | NP_006255.1 | ||||
NM_080683.2 | Intron | NP_542414.1 | ||||
NM_080684.2 | Intron | NP_542415.1 | ||||
NM_080685.2 | Intron | NP_542416.1 | ||||
XM_011532165.1 | Intron | XP_011530467.1 | ||||
XM_017008511.1 | Intron | XP_016864000.1 | ||||
XM_017008512.1 | Intron | XP_016864001.1 | ||||
XM_017008513.1 | Intron | XP_016864002.1 | ||||
XM_017008514.1 | Intron | XP_016864003.1 | ||||
XM_017008515.1 | Intron | XP_016864004.1 | ||||
XM_017008516.1 | Intron | XP_016864005.1 |
Set Membership: |
HapMap Validated |