Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCATTCCTTTTTATTGCTAAGGA[A/G]CATTTCATTGTATGGATATACCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NXPE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NXPE2 - neurexophilin and PC-esterase domain family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182495.5 | Intron | NP_872301.2 | ||||
XM_011542604.2 | Intron | XP_011540906.1 | ||||
XM_017017206.1 | Intron | XP_016872695.1 | ||||
XM_017017207.1 | Intron | XP_016872696.1 | ||||
XM_017017208.1 | Intron | XP_016872697.1 | ||||
XM_017017209.1 | Intron | XP_016872698.1 | ||||
XM_017017210.1 | Intron | XP_016872699.1 | ||||
XM_017017211.1 | Intron | XP_016872700.1 | ||||
XM_017017212.1 | Intron | XP_016872701.1 |