Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAGAGTCCCTCTTATGTGACTG[A/G]TATTCTCTGAGGGGTTTCAAACCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606029 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPSF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CPSF3 - cleavage and polyadenylation specific factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IAH1 - isoamyl acetate-hydrolyzing esterase 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039613.2 | Intron | NP_001034702.1 | ||||
NM_001320858.1 | Intron | NP_001307787.1 | ||||
NM_001320859.1 | Intron | NP_001307788.1 | ||||
NM_001320860.1 | Intron | NP_001307789.1 | ||||
NM_001320863.1 | Intron | NP_001307792.1 | ||||
XM_017003878.1 | Intron | XP_016859367.1 |
Set Membership: |
HapMap |