Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609150 MIM: 156535 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXXC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CXXC1 - CXXC finger protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101654.1 | Intron | NP_001095124.1 | ||||
NM_014593.3 | Intron | NP_055408.2 | ||||
XM_011525940.2 | Intron | XP_011524242.1 | ||||
XM_011525941.2 | Intron | XP_011524243.1 | ||||
XM_017025718.1 | Intron | XP_016881207.1 | ||||
XM_017025719.1 | Intron | XP_016881208.1 |
MBD1 - methyl-CpG binding domain protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |