Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACTGAAAATGAACAACTAAATAA[C/T]TTTGAAAATAAAGTAACTATAAACC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
MCF2L2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
MCF2L2 - MCF.2 cell line derived transforming sequence-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015078.3 | 3873 | UTR 3 | NP_055893.3 | |||
XM_011512585.2 | 3873 | UTR 3 | XP_011510887.1 | |||
XM_011512586.2 | 3873 | UTR 3 | XP_011510888.1 | |||
XM_017005943.1 | 3873 | Intron | XP_016861432.1 | |||
XM_017005944.1 | 3873 | Intron | XP_016861433.1 | |||
XM_017005945.1 | 3873 | Intron | XP_016861434.1 | |||
XM_017005946.1 | 3873 | Intron | XP_016861435.1 |