Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604915 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K13 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAP3K13 - mitogen-activated protein kinase kinase kinase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242314.1 | 5666 | UTR 3 | NP_001229243.1 | |||
NM_001242317.1 | 5666 | UTR 3 | NP_001229246.1 | |||
NM_004721.4 | 5666 | UTR 3 | NP_004712.1 | |||
XM_011513310.2 | 5666 | Intron | XP_011511612.1 | |||
XM_017007456.1 | 5666 | UTR 3 | XP_016862945.1 | |||
XM_017007457.1 | 5666 | Intron | XP_016862946.1 | |||
XM_017007458.1 | 5666 | Intron | XP_016862947.1 | |||
XM_017007459.1 | 5666 | Intron | XP_016862948.1 |
TMEM41A - transmembrane protein 41A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080652.3 | 5666 | Intron | NP_542383.1 | |||
XM_017007437.1 | 5666 | Intron | XP_016862926.1 |
Set Membership: |
HapMap |