Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605652 MIM: 609784 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FBXL2 - F-box and leucine rich repeat protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171713.1 | Intron | NP_001165184.1 | ||||
NM_012157.3 | Intron | NP_036289.3 | ||||
XM_005265014.2 | Intron | XP_005265071.1 | ||||
XM_005265015.3 | Intron | XP_005265072.1 | ||||
XM_005265016.2 | Intron | XP_005265073.1 | ||||
XM_005265017.3 | Intron | XP_005265074.1 | ||||
XM_011533559.2 | Intron | XP_011531861.1 | ||||
XM_017006092.1 | Intron | XP_016861581.1 | ||||
XM_017006093.1 | Intron | XP_016861582.1 | ||||
XM_017006094.1 | Intron | XP_016861583.1 |
UBP1 - upstream binding protein 1 (LBP-1a) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128160.1 | Intron | NP_001121632.1 | ||||
NM_001128161.1 | Intron | NP_001121633.1 | ||||
NM_014517.4 | Intron | NP_055332.3 |
Set Membership: |
HapMap Validated |