Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180073 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE6G PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PDE6G - phosphodiesterase 6G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002602.3 | 453 | UTR 3 | NP_002593.1 | |||
XM_006721942.3 | 453 | UTR 3 | XP_006722005.1 | |||
XM_017024734.1 | 453 | UTR 3 | XP_016880223.1 | |||
XM_017024735.1 | 453 | UTR 3 | XP_016880224.1 | |||
XM_017024736.1 | 453 | UTR 3 | XP_016880225.1 |
TSPAN10 - tetraspanin 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |