Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTTGCTCTTCTCACAGGTCAGC[A/G]GCCGGCAGAGCTGCTTCTGGCTTTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102680 MIM: 610977 MIM: 611526 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ADD1 - adducin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MFSD10 - major facilitator superfamily domain containing 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOP14 - NOP14 nucleolar protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291978.1 | 2442 | Missense Mutation | CCG,CTG | P,L 752 | NP_001278907.1 | |
NM_001291979.1 | 2442 | Missense Mutation | CCG,CTG | P,L 752 | NP_001278908.1 | |
NM_003703.2 | 2442 | Missense Mutation | CCG,CTG | P,L 752 | NP_003694.1 |
NOP14-AS1 - NOP14 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |