Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610284 MIM: 607166 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LIPT1 - lipoyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204830.1 | 809 | Silent Mutation | GCC,GCT | A,A 237 | NP_001191759.1 | |
NM_015929.3 | 809 | Silent Mutation | GCC,GCT | A,A 237 | NP_057013.1 | |
NM_145197.2 | 809 | Silent Mutation | GCC,GCT | A,A 237 | NP_660198.1 | |
NM_145198.2 | 809 | Silent Mutation | GCC,GCT | A,A 237 | NP_660199.1 | |
NM_145199.2 | 809 | Silent Mutation | GCC,GCT | A,A 237 | NP_660200.1 |
MITD1 - microtubule interacting and trafficking domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320417.1 | 809 | Intron | NP_001307346.1 | |||
NM_001320418.1 | 809 | Intron | NP_001307347.1 | |||
NM_001320419.1 | 809 | Intron | NP_001307348.1 | |||
NM_138798.2 | 809 | Intron | NP_620153.1 | |||
XM_011510581.2 | 809 | Intron | XP_011508883.1 | |||
XM_011510582.2 | 809 | Intron | XP_011508884.1 | |||
XM_011510583.1 | 809 | Intron | XP_011508885.1 | |||
XM_017003314.1 | 809 | Intron | XP_016858803.1 | |||
XM_017003315.1 | 809 | Intron | XP_016858804.1 |
TSGA10 - testis specific 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |