Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACATGCTTTCTTCGTTTGGGTAG[A/G]GATGGGGATCCAAGTCCCCTATGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188411 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD1E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD1E - CD1e molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042583.2 | 31 | Intron | NP_001036048.1 | |||
NM_001042584.2 | 31 | Intron | NP_001036049.1 | |||
NM_001042585.2 | 31 | Intron | NP_001036050.1 | |||
NM_001042586.2 | 31 | Intron | NP_001036051.1 | |||
NM_001042587.2 | 31 | Intron | NP_001036052.1 | |||
NM_001185107.1 | 31 | Intron | NP_001172036.1 | |||
NM_001185108.1 | 31 | Intron | NP_001172037.1 | |||
NM_001185110.1 | 31 | Intron | NP_001172039.1 | |||
NM_001185112.1 | 31 | Intron | NP_001172041.1 | |||
NM_001185113.1 | 31 | Intron | NP_001172042.1 | |||
NM_001185114.1 | 31 | Intron | NP_001172043.1 | |||
NM_001185115.1 | 31 | Intron | NP_001172044.1 | |||
NM_030893.3 | 31 | Intron | NP_112155.2 | |||
XM_011510133.2 | 31 | UTR 5 | XP_011508435.1 | |||
XM_011510134.2 | 31 | UTR 5 | XP_011508436.1 |