Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATTCCAGTTGAACAGAATAAAGG[C/T]CAAAATAGAGCTGCCTTGGGGGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 166490 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPP1 - secreted phosphoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000582.2 | 623 | Intron | NP_000573.1 | |||
NM_001040058.1 | 623 | Intron | NP_001035147.1 | |||
NM_001040060.1 | 623 | Intron | NP_001035149.1 | |||
NM_001251829.1 | 623 | Intron | NP_001238758.1 | |||
NM_001251830.1 | 623 | Missense Mutation | GCC,GTC | A,V 24 | NP_001238759.1 | |
XM_017008564.1 | 623 | Missense Mutation | GCC,GTC | A,V 24 | XP_016864053.1 |
Set Membership: |
HapMap |