Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612283 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4783 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR4783 - microRNA 4783 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PROC - protein C, inactivator of coagulation factors Va and VIIIa | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000312.3 | 66 | Intron | NP_000303.1 | |||
XM_005263717.4 | 66 | Intron | XP_005263774.1 | |||
XM_017004505.1 | 66 | Silent Mutation | GAC,GAT | D,D 17 | XP_016859994.1 | |
XM_017004506.1 | 66 | Intron | XP_016859995.1 |
Set Membership: |
HapMap |