Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCTATGCTTCAAAGCAATTAAAT[C/T]AAGGAGGATTCCAATTCTTTGGGCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601878 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLF10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KLF10 - Kruppel like factor 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032282.3 | 2374 | UTR 3 | NP_001027453.1 | |||
NM_005655.3 | 2374 | UTR 3 | NP_005646.1 |
LOC101927245 - vegetative cell wall protein gp1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |