Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGGTGGACACAGGGAGCAAGCT[A/C]CTTCGCTGTTCTCTGGGCTCATTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603551 MIM: 605082 MIM: 607052 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HYAL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HYAL2 - hyaluronoglucosaminidase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASSF1 - Ras association domain family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206957.1 | 1207 | UTR 3 | NP_001193886.1 | |||
NM_007182.4 | 1207 | UTR 3 | NP_009113.3 | |||
NM_170712.2 | 1207 | UTR 3 | NP_733830.1 | |||
NM_170713.2 | 1207 | UTR 3 | NP_733831.1 | |||
NM_170714.1 | 1207 | UTR 3 | NP_733832.1 | |||
XM_011533315.1 | 1207 | UTR 3 | XP_011531617.1 | |||
XM_011533316.2 | 1207 | UTR 3 | XP_011531618.1 |
RASSF1-AS1 - RASSF1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUSC2 - tumor suppressor candidate 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |