Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTCATGATCAGAGCCAGCTGAT[A/G]AGAAAATAACGCATCCAAGAGGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604490 MIM: 608896 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SACS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SACS - sacsin molecular chaperone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278055.1 | 14295 | UTR 3 | NP_001264984.1 | |||
NM_014363.5 | 14295 | UTR 3 | NP_055178.3 | |||
XM_005266338.2 | 14295 | UTR 3 | XP_005266395.1 | |||
XM_011535039.2 | 14295 | UTR 3 | XP_011533341.1 | |||
XM_017020539.1 | 14295 | UTR 3 | XP_016876028.1 |
SGCG - sarcoglycan gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |