Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609559 MIM: 609557 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMKMT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CAMKMT - calmodulin-lysine N-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024766.4 | Intron | NP_079042.1 | ||||
XM_011533111.2 | Intron | XP_011531413.1 | ||||
XM_011533113.2 | Intron | XP_011531415.1 | ||||
XM_017004970.1 | Intron | XP_016860459.1 | ||||
XM_017004971.1 | Intron | XP_016860460.1 | ||||
XM_017004972.1 | Intron | XP_016860461.1 | ||||
XM_017004973.1 | Intron | XP_016860462.1 | ||||
XM_017004974.1 | Intron | XP_016860463.1 | ||||
XM_017004975.1 | Intron | XP_016860464.1 | ||||
XM_017004976.1 | Intron | XP_016860465.1 | ||||
XM_017004977.1 | Intron | XP_016860466.1 | ||||
XM_017004978.1 | Intron | XP_016860467.1 | ||||
XM_017004979.1 | Intron | XP_016860468.1 | ||||
XM_017004980.1 | Intron | XP_016860469.1 | ||||
XM_017004981.1 | Intron | XP_016860470.1 | ||||
XM_017004982.1 | Intron | XP_016860471.1 | ||||
XM_017004983.1 | Intron | XP_016860472.1 |
PREPL - prolyl endopeptidase-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |