Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCATTCATTATTATATAATACC[C/T]GCATGCAAAAGGCTACAATTTAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300576 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZDHHC15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZDHHC15 - zinc finger DHHC-type containing 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146256.1 | 5490 | UTR 3 | NP_001139728.1 | |||
NM_001146257.1 | 5490 | Intron | NP_001139729.1 | |||
NM_144969.2 | 5490 | UTR 3 | NP_659406.1 | |||
XM_006724624.3 | 5490 | Intron | XP_006724687.1 | |||
XM_017029296.1 | 5490 | Intron | XP_016884785.1 | |||
XM_017029297.1 | 5490 | Intron | XP_016884786.1 | |||
XM_017029298.1 | 5490 | Intron | XP_016884787.1 |