Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGACTTTCCCGCCAAGTCTTACCT[C/T]CCAAAGTCTATCCTCTCTTGGTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147563 MIM: 605912 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IBSP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IBSP - integrin binding sialoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MEPE - matrix extracellular phosphoglycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184694.2 | Intron | NP_001171623.1 | ||||
NM_001184695.1 | Intron | NP_001171624.1 | ||||
NM_001184696.1 | Intron | NP_001171625.1 | ||||
NM_001184697.1 | Intron | NP_001171626.1 | ||||
NM_001291183.1 | Intron | NP_001278112.1 | ||||
NM_020203.3 | Intron | NP_064588.1 | ||||
XM_006714278.2 | Intron | XP_006714341.1 |