Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGATATCTCGAACAGCAGTATAGT[C/T]TTAGGATTAGAACTTGGTTAAAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604055 MIM: 615391 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IDI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
IDI1 - isopentenyl-diphosphate delta isomerase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317955.1 | Intron | NP_001304884.1 | ||||
NM_001317956.1 | Intron | NP_001304885.1 | ||||
NM_001317957.1 | Intron | NP_001304886.1 | ||||
NM_004508.3 | Intron | NP_004499.2 | ||||
XM_011519457.1 | Intron | XP_011517759.1 | ||||
XM_017016191.1 | Intron | XP_016871680.1 |
IDI2-AS1 - IDI2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |