Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATTGATTATGGGTAATTTTCATA[C/T]CTTGTCTGCCTGAAAATAAAGTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611604 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERLIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ERLIN1 - ER lipid raft associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100626.1 | Intron | NP_001094096.1 | ||||
NM_006459.3 | Intron | NP_006450.2 | ||||
XM_005269442.3 | Intron | XP_005269499.1 | ||||
XM_017015541.1 | Intron | XP_016871030.1 |