Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCCCCTTCCCAGTGGCCAGCCA[C/T]GGTGGAATCACACAGGGCCAGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602581 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOLGA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GOLGA3 - golgin A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172557.1 | 3779 | Intron | NP_001166028.1 | |||
NM_005895.3 | 3779 | UTR 3 | NP_005886.2 | |||
XM_005266164.4 | 3779 | UTR 3 | XP_005266221.1 | |||
XM_005266167.4 | 3779 | UTR 3 | XP_005266224.1 | |||
XM_006719736.3 | 3779 | UTR 3 | XP_006719799.1 | |||
XM_006719737.3 | 3779 | UTR 3 | XP_006719800.1 | |||
XM_011534793.2 | 3779 | UTR 3 | XP_011533095.1 | |||
XM_011534794.2 | 3779 | UTR 3 | XP_011533096.1 |