Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609895 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CASZ1 - castor zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079843.2 | Intron | NP_001073312.1 | ||||
NM_017766.4 | Intron | NP_060236.3 | ||||
XM_005263479.3 | Intron | XP_005263536.1 | ||||
XM_011541635.2 | Intron | XP_011539937.1 | ||||
XM_017001539.1 | Intron | XP_016857028.1 | ||||
XM_017001540.1 | Intron | XP_016857029.1 | ||||
XM_017001541.1 | Intron | XP_016857030.1 |
Set Membership: |
Validated |